Boreal Science Compound Beginner Microscopes
Compound Coarse and Fine Focus Beginner Microscope with Gliding Stage, Cordless LED
Catalog # 470177-096
Supplier: VWR
Fournisseur #: 5791200
Jump to:
Product Details & Documents
Boreal beginner microscopes are ideal for introducing students to microscopy.
- All-Glass-Coated Optics
- All-Metal Construction
- Parfocal and Parcentered
- Objectives: 4x, 10x, and 40x
- Boreal Lifetime Warranty
It's scaled down for younger students and is simple to use, but it also prepares students for the transition to high school instruction.
Rugged metal components are engineered for years of reliable performance. Student-proof features minimize loss of parts.
Ordering information: This product includes a manufacturer's warranty.
Spécifications
- Désignation:Compound Coarse and Fine Focus Beginner Microscope with Gliding Stage
- Illumination:Cordless LED