Ward's® Merismopedia
Merismopedia (Small), Soil water medium, 30, Tube
Catalog # 470176-610
Supplier: Ward's Science
Fournisseur #: LB1902
Jump to:
Product Details & Documents
Freshwater. Colonial. Sheet of many cells one cell thick. Soil-water medium.
Shipped in a form that provides the best quality specimen. Most are supplied in 16 x 125 mm tubes, 2 oz. jars, or 8 oz. jars.
Spécifications
- Désignation:Merismopedia (Small)
- No. of Students:30
- Packaging:Tube
- Media:Soil water medium