Didinium
Didinium
Didinium Culture for 100 Students
Catalog # 470176-268
Supplier:   Avantor
Didinium
Catalog # 470176-268
Supplier:  Avantor
Didinium
Didinium
Didinium Culture for 100 Students
Price
$45.75
Log in pour la disponibilité
Each
Product Details & Documents
Predatory. Feeds on Paramecium caudatum. Hay medium.
Characterized by cilia; nearly all possess two types of nuclei.
Spécifications
  • Désignation:
    Didinium Culture for 100 Students