Didinium
Didinium Culture for 100 Students
Catalog # 470176-268
Supplier: Avantor
Jump to:
Product Details & Documents
Predatory. Feeds on Paramecium caudatum. Hay medium.
Characterized by cilia; nearly all possess two types of nuclei.
Spécifications
- Désignation:Didinium Culture for 100 Students