Didinium
Didinium (Small), Hay culture medium, 30, Jar
Catalog # 470176-266
Supplier: Ward's Science
Fournisseur #: DID30/35
Jump to:
Product Details & Documents
Predatory. Feeds on Paramecium caudatum. Hay medium.
Characterized by cilia; nearly all possess two types of nuclei.
Spécifications
- Désignation:Didinium (Small)
- No. of Students:30
- Packaging:Jar
- Media:Hay culture medium