Banana Plug Patch Cords | Avantor
Banana Plug Patch Cords
Patch cord, red, 61 cm, banana plugs both ends
Catalog # 470176-240
Fournisseur #:  1350302150002
Banana Plug Patch Cords
Catalog # 470176-240
Banana Plug Patch Cords
Banana Plug Patch Cords
Patch cord, red, 61 cm, banana plugs both ends
Price
$14.20
Log in pour la disponibilité
Each Pack of 6
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Double ended banana plug jumper wires.
  • Available in red or black
  • 15 cm (6") or 61 cm (24") long
Stackable 4 mm banana plugs on both ends of these flexible, plastic coated wires. The wires have a rating of 15 A, AC.
Spécifications
  • Désignation:
    Patch cord, red, 61 cm, banana plugs both ends