Disposable Specimen Containers
Specimen Container, 0.91 L, Disposable
Catalog # 470173-880
Supplier: Dynalon
Fournisseur #: 453615
Jump to:
Product Details & Documents
Keep specimens organized and out of your way with these handy storage units.
Featuring high-density polyethylene construction and tight-fitting lids, these translucent containers are resistant to freezing and boiling, and are sturdy enough to be used multiple times.
Spécifications
- Désignation:Specimen Container, 0.91 L, Disposable