Disposable Specimen Containers
Specimen Container, 1.81 L, Disposable
Catalog # 470173-878
Fournisseur #: 453625
Jump to:
Discontinued
Product Details & Documents
Keep specimens organized and out of your way with these handy storage units.
Featuring high-density polyethylene construction and tight-fitting lids, these translucent containers are resistant to freezing and boiling, and are sturdy enough to be used multiple times.
Spécifications
- Désignation:Specimen Container, 1.81 L, Disposable