Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Plastic, Snap-On Cap Vials

Plastic, Snap-On Cap Vials

Culture vials, 148.00 ml, 47×85 mm, Oui, Snap-on cap
Catalog # 470157-876
Supplier:   Thornton Plastics
Supplier #:  40 C
Plastic, Snap-On Cap Vials
Catalog # 470157-876
Supplier:  Thornton Plastics
Plastic, Snap-On Cap Vials
Plastic, Snap-On Cap Vials

Culture vials, 148.00 ml, 47×85 mm, Oui, Snap-on cap

Price
$25.60
Log in for availability
Pack of 12 Case of 240
Product Details & Documents

Store cultures, specimens or other items in these handy clear plastic vials.

Specifications
  • Avec bouchon:
    Oui
  • Cap Type:
    Snap-on cap
  • Capacité:
    148.00 ml
  • Désignation:
    Culture vials
  • Diameter ×H:
    47×85 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.