Precision pH Test Strips
pH paper strips, 10.0–13.0
Catalog # 470150-880
Supplier: Precision Laboratories
Fournisseur #: PH1013-1B-50
Jump to:
Product Details & Documents
The indicator on these strips won’t contaminate the solutions the strips are testing.
- Vinyl-backed
- Non-contaminating indicator
- Fifty strips per package
Durable pH testing strips that are backed with vinyl.
Spécifications
- Désignation:pH paper strips
- pH Range:10.0–13.0