Cellophane
Cellophane Roll, Blue, 20×100"
Catalog # 470150-816
Supplier: HYGLOSS PRODUCTS, INC.
Fournisseur #: 71006
Jump to:
Product Details & Documents
Use the various colors to demonstrate plants’ reaction to light waves.
- Made from recycled materials
Also useful for demonstrating the effects of mixing various colors of light.
Spécifications
- Color:Blue
- Désignation:Cellophane Roll
- Taille:20×100"