Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Sunprint Kit | Avantor

Sunprint Kit

Refill, Sunprint Kit, 12 Sheets, 10 cm x 10 cm each
Catalog # 470149-764
Fournisseur #:  W220B
Sunprint Kit
Catalog # 470149-764
Sunprint Kit
Sunprint Kit

Refill, Sunprint Kit, 12 Sheets, 10 cm x 10 cm each

Price
$7.40
Log in pour la disponibilité
Pack of 12
Product Details & Documents

The Sunprint Kit includes sun sensitive paper to help you make photographic prints using the power of the sun.

To make images, just place an array of interesting objects on a sheet of Sunprint™ paper, expose it to the sun for a few minutes, immerse in water and watch as a permanent image appears.

Delivery information: Includes 12 sheets of sun-sensitive paper (10×10cm), acrylic plate and instructions.

Spécifications
  • Désignation:
    Refill, Sunprint Kit, 12 Sheets, 10 cm x 10 cm each
Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.