$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
PAN ALUMINUM FOIL 12X8.6X1.2IN 31X22X3CM
470149-622
:
REGIONAL DISTRIBUTORS, INC.
PAN ALUMINUM FOIL 12X8.6X1.2IN 31X22X3CM
470149-622
:
REGIONAL DISTRIBUTORS, INC.
:
22931
Frequently Bought Together
Méthanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
: MilliporeSigma
VWR® Disposable Serological Pipettes
: VWR International
VWR® Universal Pipette Tips
: VWR International
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
: MilliporeSigma
VWR® Nitrile Examination Gloves, Blue
: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
: Corning
VWR® Disposable Petri Dishes
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH