Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Jars and Caps | Avantor

Jars and Caps

89 mm, PET jar, 16 oz.
Catalog # 470147-072
Fournisseur #:  RM89WHT024
Jars and Caps
Catalog # 470147-072
Jars and Caps
Jars and Caps

89 mm, PET jar, 16 oz.

Price
$6.25
Log in pour la disponibilité
Each Case of 350
Product Details & Documents
Spécifications
  • Capacité:
    16 oz.
  • Désignation:
    89 mm, PET jar
  • Cat. No.:
    470147-072
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.