Boreal Science Quieting Solution
Boreal Quieting Solution, 30 mL, For Protozoan Slowing
Catalog # 470106-046
Supplier:   Ward's Science
 :  BB0161-30M
Boreal Science Quieting Solution
Catalog # 470106-046
Supplier:  Ward's Science
CAS Number:  
Boreal Science Quieting Solution
Boreal Science Quieting Solution
Boreal Quieting Solution, 30 mL, For Protozoan Slowing
CAS Number:  
Price
$14.75
Log in pour la disponibilité
Each
Product Details & Documents
Use this solution to slow down and relax microorganisms such as Paramecium, Euglena and Stentor. The ready-to-use dropper bottle makes it easy to store and transfer quieting liquid. Includes a dropper.
Please Note: This product is designed for educational and teaching laboratories and no certificate of analysis is available.
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Spécifications
Spécifications
  • Désignation:
    Boreal Quieting Solution, 30 mL, For Protozoan Slowing
Spécifications