Pulleys
Pulley, Double Sheave
Catalog # 470104-442
Supplier: Wards
Fournisseur #: PH0280B
Jump to:
Product Details & Documents
These free-running plastic pulleys are durable in design.
- Plastic Construction
- Durable
- Nickel-Plated Steel Blocks with Hooks
The sheaves are 50 mm in Ø and made of rigid plastic.
The nickel-plated steel blocks have hooks at the top and bottom.
Spécifications
- Désignation:Pulley, Double Sheave