Friction Rods | Avantor
Friction Rods
Hard Nylon, Flat Ended
Catalog # 470104-254
Supplier:   CURBELL PLASTICS
Fournisseur #:  BLNYLROD
Friction Rods
Catalog # 470104-254
Supplier:  CURBELL PLASTICS
Friction Rods
Friction Rods
Hard Nylon, Flat Ended
Price
$34.30
Log in pour la disponibilité
Each
Product Details & Documents
For your Electrostatic Needs!
  • Excellent quality
Friction rods are used for producing static charges when rubbed against materials such as wool, flannel, silk, and fur.
Spécifications
  • Désignation:
    Hard Nylon, Flat Ended