Balls
Golf ball, 1.76", -
470092-592
: 2931400
Useful for collision and pendulum experiments.
A must have in any physics classroom or laboratory.
- Désignation:Golf ball
- Packaging:-
- Diameter:1.76"
Useful for collision and pendulum experiments.
A must have in any physics classroom or laboratory.