Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Glucose Test Strips | Avantor

Glucose Test Strips

Glucose Test Strips
Catalog # 470089-552
Fournisseur #:  GLU-1B-50
Glucose Test Strips
Catalog # 470089-552
CAS Number:  
Glucose Test Strips
Glucose Test Strips

Glucose Test Strips

CAS Number:  
Price
37,85 $
Log in pour la disponibilité
Each
Product Details & Documents

Storage: Green

Please Note: This product is designed for educational and teaching laboratories and no certificate of analysis is available.

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Spécifications

Spécifications

  • Désignation:
    Glucose Test Strips

Spécifications