Solid Balls for Physics Experiments
Solid Balls for Physics Experiments
Solid Steel Ball, 10 mm, ³/₈", -
  470089-322
Supplier #:  2754800
Solid Balls for Physics Experiments
  470089-322
Solid Balls for Physics Experiments
Solid Balls for Physics Experiments
Solid Steel Ball, 10 mm, ³/₈", -
$4.35
Each
Drilled balls are used for pendulum bobs or impact experiments, while our other balls are suitable for collision apparatus and density/volume experiments.
Caution: WARNING: CHOKING HAZARD -Item contains a small ball. Not for children under 3 years.
  • Désignation:
    Solid Steel Ball, 10 mm
  • Packaging:
    -
  • Diameter:
    ³/₈"