Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Straws, Plastic | Avantor

Straws, Plastic

Straws, Unwrapped, 0.73 cm, 19.7 cm, Clear
Catalog # 470089-218
Supplier:   Wards
Fournisseur #:  1513 CLEAR
Straws, Plastic
Catalog # 470089-218
Supplier:  Wards
Straws, Plastic
Straws, Plastic

Straws, Unwrapped, 0.73 cm, 19.7 cm, Clear

Price
$10.00
Log in pour la disponibilité
Pack of 1
Product Details & Documents

Straws made of plastic.

Spécifications
  • Color:
    Clear
  • Désignation:
    Straws, Unwrapped
  • Longueur:
    19.7 cm
  • Diameter:
    0.73 cm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.