Cylinder Guards
Cylinder Guards
Cylinder Guards, For Cylinder Cap., 100 mL
Catalog # 470151-204
Supplier #:  VCYCG3
Cylinder Guards
Catalog # 470151-204
Supplier:  
Cylinder Guards
Cylinder Guards
Cylinder Guards, For Cylinder Cap., 100 mL
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
VWR GUARD CYL SPNG-RBR 50/100ML PK12 Catalog #15001-378
Product 15001-378 is similar to 470151-204 but manufacturer and specifications may vary.
Product Details & Documents
Black sponge rubber guards prevent breakage and rolling.
  • Hexagonal
  • 13 mm thick
Specifications
  • Désignation:
    Cylinder Guards, For Cylinder Cap., 100 mL