Microscope Immersion Oil, Cargille
Product Details & Documents
Types A and B are virtually interchangeable and are miscible with each other for intermediate viscosities. Produced in larger quantities than other types. Type A, at 150 centistokes, reduces any tendancy to trap air. Type B, at 1250 centistokes, is thick enough for viewing multiple slides with one application.
Types A and B are virtually interchangeable and are miscible with each other for intermediate viscosities
Type NVH is used for inverted, inclined, projection, and long focus instruments. The greater the gap between the cover glass and objective, or between the slide and condenser, the more desirable high viscosity becomes. The very high viscosities of Type NVH at 21,000 cSt give excellent results for these applications.
Type 300 is designed for use with automated hematology systems for successful imaging and mechanical processing. Type 300 is manufactured to meet the stringent requirements of this equipment, which include specialized viscosity and exacting controls for consistency.
Produced in larger quantities than other types. Type A, at 150 centistokes, reduces any tendancy to trap air. Type B, at 1250 centistokes, is thick enough for viewing multiple slides with one application.
Spécifications
Spécifications
- Désignation:Type A Microscope Immersion Oil
- Taille:118 ml (4 oz.)