Carbon Rod Electrode
Carbon Rod Electrode, Laboratory, Pack of 12
Catalog # 470149-520
Supplier: Shiv Dial Sud & Sons
Supplier #: PH50067/4
Jump to:
Product Details & Documents
CAS Number: 7440-44-0
Formula Weight: 12.01
Formula: C
Shelf Life (months): 36
Storage: Green
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Specifications
Specifications
- Désignation:Carbon Rod Electrode
- Taille:Pack of 12
- Chemical/Physical Specifications:6×125 mm
- Grade:Laboratory



