Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
VWR® Tall Form Beaker | Avantor

VWR® Tall Form Beaker

1000 ml, 50 ml, 50 - 950 ml
Catalog # 470149-346
Supplier:   VWR International
VWR® Tall Form Beaker
Catalog # 470149-346
Supplier:  VWR International
VWR® Tall Form Beaker

1000 ml, 50 ml, 50 - 950 ml

Price
$185.20
Log in pour la disponibilité
Pack of 6 Case of 18
Product Details & Documents

High Quality at a Great Value

  • Borosilicate Glass Fabrication
  • Graduated Markings
  • Easy-to-Measure
  • Beakers Meet ASTM Standards

Documents

Spécifications
  • Intervalle de graduation:
    50 ml
  • Plage de graduation:
    50 - 950 ml
  • Capacité:
    1000 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.