Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
VWR® Tall Form Beaker | Avantor

VWR® Tall Form Beaker

300 ml, 25 ml, 25 - 250 ml
Catalog # 470149-328
Supplier:   VWR International
VWR® Tall Form Beaker
Catalog # 470149-328
Supplier:  VWR International
VWR® Tall Form Beaker

300 ml, 25 ml, 25 - 250 ml

Price
$164.30
Log in pour la disponibilité
Pack of 12 Case of 48
Product Details & Documents

High Quality at a Great Value

  • Borosilicate Glass Fabrication
  • Graduated Markings
  • Easy-to-Measure
  • Beakers Meet ASTM Standards

Documents

Spécifications
  • Intervalle de graduation:
    25 ml
  • Plage de graduation:
    25 - 250 ml
  • Capacité:
    300 ml