Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
VWR® Tall Form Beaker | Avantor

VWR® Tall Form Beaker

500 ml, 50 ml, 50 - 450 ml
Catalog # 470149-326
Supplier:   VWR International
VWR® Tall Form Beaker
Catalog # 470149-326
Supplier:  VWR International
VWR® Tall Form Beaker

500 ml, 50 ml, 50 - 450 ml

Price
$91.15
Log in pour la disponibilité
Pack of 6 Case of 30
Product Details & Documents

High Quality at a Great Value

  • Borosilicate Glass Fabrication
  • Graduated Markings
  • Easy-to-Measure
  • Beakers Meet ASTM Standards

Documents

Spécifications
  • Intervalle de graduation:
    50 ml
  • Plage de graduation:
    50 - 450 ml
  • Capacité:
    500 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.