VWR® Florence Boiling Flasks
1000 ml, 8
Supplier: VWR International
Jump to:
Product Details & Documents
High Quality Borosilicate Glass
- Meet ASTM standards
- Round body and long neck are ideal for boiling liquids without excessively rapid evaporation
Ordering information: Rubber stoppers are not included.
Documents
Certificates
Search by Lot NumberSpecifications
- Capacité:1000 ml
- Rubber Stopper Size:8