VWR® Test Tubes
14 ml, 00, 15×125 mm
Supplier: VWR International
Jump to:
Product Details & Documents
VWR® Test tubes are made from high-quality glass.
- Borosilicate glass construction
- Uniform thickness
Each tube features a strong lip.
Certifications: ASTM standards.
Documents
Certificates
Recherche par numéro de lotSpécifications
- Capacité:14 ml
- Stopper Size:00
- O.D.×L:15×125 mm