Bare Nichrome Wire
Bare Nichrome Wire
Bare Nichrome wire, 18 SWG
Catalog # 470149-230
Supplier:   Wards
Supplier #:  PH1064A
Bare Nichrome Wire
Catalog # 470149-230
Supplier:  Wards
Bare Nichrome Wire
Bare Nichrome Wire
Bare Nichrome wire, 18 SWG
Price
$67.75
Log in for availability
Each Pack of 10
Product Details & Documents
Create High Performance Loops
125gm reels of bare Nichrome wire for use in resistance heating or inoculating loops.
Specifications
  • Désignation:
    Bare Nichrome wire
  • Jauge:
    18 SWG