Bare Nichrome Wire | Avantor
Bare Nichrome Wire
Bare Nichrome wire, 20 SWG
Catalog # 470149-228
Supplier:   Wards
Fournisseur #:  PH1064B
Bare Nichrome Wire
Catalog # 470149-228
Supplier:  Wards
Bare Nichrome Wire
Bare Nichrome Wire
Bare Nichrome wire, 20 SWG
Liquidation
$23.85
Vente
Valable jusqu'au 31/12/2026
Log in pour la disponibilité
Each Pack of 10
Product Details & Documents
Create High Performance Loops
125gm reels of bare Nichrome wire for use in resistance heating or inoculating loops.
Spécifications
  • Désignation:
    Bare Nichrome wire
  • Jauge:
    20 SWG