Bare Nichrome Wire
Bare Nichrome wire, 20 SWG
Catalog # 470149-228
Supplier: Wards
Fournisseur #: PH1064B
Jump to:
Product Details & Documents
Create High Performance Loops
125gm reels of bare Nichrome wire for use in resistance heating or inoculating loops.
Spécifications
- Désignation:Bare Nichrome wire
- Jauge:20 SWG