Bare Nichrome Wire
Bare Nichrome wire, 22 SWG
Catalog # 470149-226
Supplier: Wards
Fournisseur #: PH1064C
Jump to:
Product Details & Documents
Create High Performance Loops
125gm reels of bare Nichrome wire for use in resistance heating or inoculating loops.
Spécifications
- Désignation:Bare Nichrome wire
- Jauge:22 SWG