Disposable Micropipet Tips
0.5 - 10 µl, Naturel, 1000 Tips/Bag
Catalog # 470149-174
Supplier: VWR International
Jump to:
Product Details & Documents
High quality micropipette tips to meet any lab's needs.
- Certified DNase, RNase and pyrogen free
- Material: PP
- Autoclavable
- Designed to fit most standard micropipettes
All quantities are supplied in bulk bags.
Spécifications
- Volume:0.5 - 10 µl
- Tip Color:Naturel
- Packed:1000 Tips/Bag