VWR® Volumetric Flasks
100 ml, Glass Stopper
Supplier: VWR International
Jump to:
Product Details & Documents
These Class B borosilicate volumetric flasks feature capacity tolerances twice those specified for Class A volumetric ware.
- Class B borosilicate glass
- Includes choice of glass stopper or snap cap
- Graduation line for accurate measurements
Documents
Certificates
Recherche par numéro de lotSpécifications
- Capacité:100 ml
- Stopper Type:Glass Stopper