VWR® Volumetric Flasks
250 ml
Supplier: VWR International
Jump to:
Product Details & Documents
Our borosilicate flasks meet ASTM standards.
- Class A Flasks
- Borosilicate Glass Construction
- Graduated Marking for Measurements
- Economical
- Snap-Caps Included
The necks fit the included snap-caps.
The Class A, volumetric flasks include a graduation line for accurate measurements.
Documents
Certificates
Recherche par numéro de lotSpécifications
- Capacité:250 ml