Color-Coded Inoculating Loops, Dynalon
Color-Coded Inoculating Loops, Clear, w/0.1 mm dia. loop
470147-478
: Dynalon
: 226515
Made of color-coded styrene, these inoculating loops are easy to organize.
- Two Sizes
- Color-Coded Styrene to Distinguish Loop Size
- Sturdy, 7½" Handles
- Twenty Loops per Package
Both sizes of loops have sturdy, 7½" handles.
The handles and color-coding make these loops convenient and simple to use.
: One package has 20 loops.
- Désignation:Color-Coded Inoculating Loops, Clear, w/0.1 mm dia. loop