Polypropylene Jars
Polypropylene Jars, 125 ml
Catalog # 470147-470
Supplier: Dynalon
Fournisseur #: 421135
Jump to:
Product Details & Documents
Convenient for Storing and Collecting Lab Supplies and Specimens.
- Supplied with linerless polypropylene screw caps
- Leak-proof
- Autoclavable
Spécifications
- Capacité:125 ml
- Désignation:Polypropylene Jars