Illustrated Dissection Guides
Pigeon Guide, 16, 22 cm, 14 cm
Catalog # 470146-796
Supplier: Ward's Science
Supplier #: 420.4124.1
Jump to:
Product Details & Documents
These economical and concise guides contain the right amount of information for reference during dissection.
- Basic Facts on Specimens
- Convenient Size
- Paperback
- Color Illustrations
- Step-by-Step Instructions
Each booklet provides basic facts on specimens, step-by-step dissection instructions, and clear illustrations to guide students as they dissect their own specimens.
The paperback guides are small, so students can have them whenever they're needed.
Specifications
- Désignation:Pigeon Guide
- Longueur:22 cm
- No. of Pages:16
- Width:14 cm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



