PYREX® Burets
Geissler buret replacement tip, pack of 12, six grinded tips and six clear (ungrinded) tipss
Catalog # 470146-252
Supplier: Corning
Supplier #: 2116-GTO2
Jump to:
Product Details & Documents
Educational grade Pyrex burets.
- Heavy duty
- Transparent
Supplied with self-lubricating, leakproof PMP stopcock. This buret is unaffected by most dilute aqueous solutions, except ammonia and HF. It is not recommended for use with ethanol or organic solvents. The clear white markings make dark liquids easier to measure the the Teflon stopcock is less likely to freeze up.
Spécifications
- Désignation:Geissler buret replacement tip, pack of 12, six grinded tips and six clear (ungrinded) tipss
- Cat. No.:470146-252
- Environmental certification:I-RECs, ISO 14001 - Monterrey
- Environmentally Preferable:Low Manufacturing Impact