Single Factor Water Test Kits | Avantor
Single Factor Water Test Kits
Single factor water test kit, iron, 30 tests
Catalog # 470144-534
Fournisseur #:  480125
Single Factor Water Test Kits
Catalog # 470144-534
Single Factor Water Test Kits
Single Factor Water Test Kits
Single factor water test kit, iron, 30 tests
Price
$58.40
Log in pour la disponibilité
Each Case of 12
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Simply Dip in Water and Read the Results
These kits feature a wide range of test factors for a fraction of the price of other single factor test kits. These test kits also eliminate the need for liquid reagents, hazardous chemicals, and glassware. Each kit comes with instructions.
Spécifications
  • Désignation:
    Single factor water test kit, iron, 30 tests