Boss Head, Double Large | Avantor
Boss Head, Double Large
Boss Head Double Large, Black Coated
Catalog # 470137-840
Supplier:   Wards
Fournisseur #:  CH0665B
Boss Head, Double Large
Catalog # 470137-840
Supplier:  Wards
Boss Head, Double Large
Boss Head, Double Large
Boss Head Double Large, Black Coated
Price
$26.95
Log in pour la disponibilité
Each
Product Details & Documents
Lab Aid, Many Uses
This versatile boss head can hold rods ranging from 5-20mm in diameter. Choose from chrome-plated or black finish. The die cast boss head is fitted with plastic cap screws for easy tightening.
Spécifications
  • Désignation:
    Boss Head Double Large, Black Coated