Boss Head, Double Large
Boss Head Double Large, Black Coated
Catalog # 470137-840
Supplier: Wards
Fournisseur #: CH0665B
Jump to:
Product Details & Documents
Lab Aid, Many Uses
This versatile boss head can hold rods ranging from 5-20mm in diameter. Choose from chrome-plated or black finish. The die cast boss head is fitted with plastic cap screws for easy tightening.
Spécifications
- Désignation:Boss Head Double Large, Black Coated