Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Balls for Physics Experiments | Avantor

Balls for Physics Experiments

Drilled Copper Ball, 25 mm, 1"
Catalog # 470137-642
Supplier:   Wards
Fournisseur #:  PH0306DR-25
Balls for Physics Experiments
Catalog # 470137-642
Supplier:  Wards
Balls for Physics Experiments
Balls for Physics Experiments

Drilled Copper Ball, 25 mm, 1"

Liquidation
$18.65
Vente

Valable jusqu'au 31/12/2026

Log in pour la disponibilité
Each
Product Details & Documents

Ideal for Collision and Pendulum Demonstrations

Drilled balls for pendulum bobs or impact experiments. 

Spécifications
  • Désignation:
    Drilled Copper Ball, 25 mm
  • Diameter:
    1"
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.