Clearview Classroom Stream Table
Clearview Classroom Stream Table
Clearview Classroom Stream Table, 20", 48"
Catalog # 470136-880
Supplier #:  651-6215
Clearview Classroom Stream Table
Catalog # 470136-880
Supplier:  
Clearview Classroom Stream Table
Clearview Classroom Stream Table
Clearview Classroom Stream Table, 20", 48"
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
WARD'S DELUXE LAND SIM SYS W/CART (REV.) Catalog #470189-702
Product 470189-702 is similar to 470136-880 but manufacturer and specifications may vary.
Product Details & Documents
Simulate a flowing stream and study erosion with this classroom stream table.
  • Clear surface for visual study
    Ideal for environmental and water studies
    Water tight
    Easy to store
Bring Water Dynamics to Life with the Clearview Classroom Stream Table - a must have for any earth science classroom! This interactive stream table offers a dynamic way for students to explore erosion, sediment transport, and river formation. Designed for easy setup and durability, the rugged ABS construction features an all-in-one system—just add water, activate the pump, and start experimenting. Five adjustable water jets simulate natural water flow, from small streams to converging trib. Ships oversize.
Spécifications
  • Désignation:
    Clearview Classroom Stream Table
  • Longueur:
    48"
  • Width:
    20"