Convex Flask Brushes
Convex Flask Brushes, Hog hair, White, Plastic, 305 mm
Catalog # 470123-990
Supplier: Mill-Rose
Supplier #: 2130
Jump to:
Product Details & Documents
White hog bristles mounted on a durable, plastic handle. Flexible brush section adapts to curvature of flasks.
Specifications
- Bristle Color:White
- Bristle Material:Hog hair
- Handle Material:Plastic
- Désignation:Convex Flask Brushes
- Longueur totale:305 mm