Squeeze-Jet Bottles
Squeeze-Jet Bottles, 30 ml
Catalog # 470123-920
Supplier: Dynalon
Supplier #: 524165-0030
Jump to:
Product Details & Documents
Clear Polyethylene.
- Use for drop dispensing or for a jet spray
- Air-tight seal is made when the dispenser nozzle is closed
Specifications
- Capacité:30 ml
- Désignation:Squeeze-Jet Bottles