Gas Generating Bottle Set | Avantor
Gas Generating Bottle Set
Gas Generating Bottle Set, Capacity: 500 mL
Catalog # 470123-890
Supplier:   GSC International
Fournisseur #:  9001-1
Gas Generating Bottle Set
Catalog # 470123-890
Supplier:  GSC International
Gas Generating Bottle Set
Gas Generating Bottle Set
Gas Generating Bottle Set, Capacity: 500 mL
Price
32,25 $
Log in pour la disponibilité
Each
Product Details & Documents
When used with a pneumatic trough, our gas generating bottle set is ideal for generating small quantities of hydrogen and other gases.
  • Hydrogen gas production requires 3M HCl and mossy zinc
The set consists of a bottle, 2-hole rubber stopper, reagent addition (thistle) tube, and gas delivery tube.
Ordering information: Pneumatic trough, ¹/₄" Int.Ø rubber tubing, and gas collection bottles are sold separately.
Spécifications
  • Désignation:
    Gas Generating Bottle Set, Capacity: 500 mL