Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Orbital Model Sets

Orbital Model Sets

Set of 5 d Orbitals: dx2, dxy, dxz, dzx, d(x2-y2)
Catalog # 470122-988
Supplier:   Klinger
Supplier #:  0606K + PKG
Orbital Model Sets
Catalog # 470122-988
Supplier:  Klinger
Orbital Model Sets
Orbital Model Sets

Set of 5 d Orbitals: dx2, dxy, dxz, dzx, d(x2-y2)

Price
$1,152.65
Log in for availability
Kit of 1
Product Details & Documents

Allow Students to Visually See Electron Orbitals

  • Completely Assembled
  • Each Model 10" High

Angular distribution of the s, p, d and f orbitals is illustrated by durable, colorful, three-dimensional models. Ideal additions for any chemistry classroom, models are carefully made of finished wood and mounted on heavy wire coordinate frames. Positive lobes, spheres, disks and crinolines are painted in red enamel; negative parts are yellow. Each model measures 10" high and is supported on a circular hardwood base. Choose from the sets listed below. Furnished completely assembled with background information. Designed by Dr. A.H. Norbury, Lecturer, Dept. of Chemistry, Loughborough University, England.

Specifications
  • Désignation:
    Set of 5 d Orbitals: dx2, dxy, dxz, dzx, d(x2-y2)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.