Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Electric Heaters | Avantor

Electric Heaters

Electric Heaters, Power, watts: 200, Total Length: 6", Heated Length: 3"
Catalog # 470122-464
Supplier:   Glo-Quartz
Fournisseur #:  LHP-200
Electric Heaters
Catalog # 470122-464
Supplier:  Glo-Quartz
Electric Heaters

Electric Heaters, Power, watts: 200, Total Length: 6", Heated Length: 3"

Price
$228.40
Log in pour la disponibilité
Each
Product Details & Documents

Excellent for heating all acid solutions except hydrofluoric.

  • Resistance element is completely sealed in a quartz tube
  • Upper portion remains cool and can be held in the bare hand
  • Diameter: ⁵/₁₆"

Permanently attached 6' 2-wire line cord/plug for 115 VAC or DC.

Spécifications
  • Désignation:
    Electric Heaters, Power, watts: 200, Total Length: 6", Heated Length: 3"