Electric Heaters
470122-462
Excellent for heating all acid solutions except hydrofluoric.
- Resistance element is completely sealed in a quartz tube
- Upper portion remains cool and can be held in the bare hand
- Diameter: ⁵/₁₆"
Permanently attached 6' 2-wire line cord/plug for 115 VAC or DC.