Hanna Instruments pH Electrode Storage Solution (230 ml)
pH Electrode Storage Solution
  470122-066
 :  HI70300M
Hanna Instruments pH Electrode Storage Solution (230 ml)
  470122-066
 :  
 :  
Hanna Instruments pH Electrode Storage Solution (230 ml)
Hanna Instruments pH Electrode Storage Solution (230 ml)
pH Electrode Storage Solution
 :  
This electrode storage solution prepared with reagent grade chemicals that can be used to ensure optimum performance of your pH and ORP electrodes. Properly storing your pH electrode in a solution keeps the glass membrane well hydrated which maintains proper function and provides accurate readings.
  • Special formulation to minimize bacterial growth on your electrode
  • Designed for storing any pH or ORP electrode
  • Air tight bottle with a tamper-proof seal Ensures quality and freshness of solution
This storage solution is a specially formulated electrode storage solution that can be used to store your pH electrode*. To ensure the best response time, the sensor tip and the reference junction of the pH electrode should be kept moist when not in use. Placing the pH electrode in a small glass filled with storage solution or replacing the solution in the protective cap is a n easy way to store the electrode. This storage solution should also be used to rehydrate the electrode after cleaning by soaking for at least one hour before taking measurements.
  • Désignation:
    pH Electrode Storage Solution