Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

C-Clamps

C-Clamp, 3.25" Depth, 6"
Catalog # 470121-796
Supplier:   GSC International
 :  C-CLAMP6
C-Clamps
Catalog # 470121-796
Supplier:  GSC International
C-Clamps
C-Clamps

C-Clamp, 3.25" Depth, 6"

Price
24,15 $
Log in pour la disponibilité
Each
Product Details & Documents

Secure Items Firmly to A Table or Workbench.

  • Natural malleable finish
  • Corrosion resistant
  • Sliding vise-type handles
Spécifications
  • Désignation:
    C-Clamp, 3.25" Depth
  • Jaw Size:
    6"
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.